(S)-2-(3-Acetamidopropanamido)-3-(1H-imidazol-4-yl)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Acetylcarnosine
A317466-1G
A317466-25G
A317466-5G
A317466-10G
A317466-250MG
103825-134EA
25
USD
103825-134
103836-066
76838-916
76838-918
76838-920
77114-680
77685-900
(S)-2-(3-Acetamidopropanamido)-3-(1H-imidazol-4-yl)propanoic acid 97%
(S)-2-(3-Acetamidopropanamido)-3-(1H-imidazol-4-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₆N₄O₄ MW: 268.27 g/mol Boiling Pt: 775.854 °C (760 mm Hg) Flash Pt: 423.021 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039376 CAS Number: 56353-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...