Glycyl-L-leucine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-Aminoacetamido)-4-methylpentanoic acid, H-Gly-Leu-OH

A274054-100G A274054-1G A274054-25G A274054-5G
103813-178EA 9 USD
103813-178 103853-102 77194-036 77305-158 77305-160 77305-162
Glycyl-L-leucine 97%
Glycyl-L-leucine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
Boiling Pt: 410.4 °C (760 mm Hg)
Melting Pt: 233…235 °C
Flash Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008127
CAS Number: 869-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR