(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, Boc-β-HomoPro-OH

A155061-100MG A155061-10G A155061-250MG A155061-1G A155061-25G A155061-5G
103804-710EA 85 USD
103804-710 103842-906 76973-270 76973-272 76973-274 77300-098 77300-100 77300-102
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 98%
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Boiling Pt: 357.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01862939
CAS Number: 56502-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR