(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid 97%

Supplier: Ambeed
Warning

A157534-10G A157534-250MG A157534-1G
103805-014EA 18 USD
103805-014 103843-214 77110-546 77110-548 77319-044
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid 97%
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.2729 g/mol
Boiling Pt: 357.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD05861546
CAS Number: 204688-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR