(S)-Glycidyl trityl ether 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane, (S)-2-((Trityloxy)methyl)oxirane

A122374-100G A122374-10G A122374-25G A122374-1G
103800-214EA 5 USD
103800-214 76718-594 76718-596 76718-598 77312-348
(S)-Glycidyl trityl ether 98%
(S)-Glycidyl trityl ether

Precaución: for Research Use Only

Formula: C₂₂H₂₀O₂
MW: 316.40 g/mol
Boiling Pt: 438.8 °C (760 mm Hg)
Melting Pt: 98…101 °C
Flash Pt: 164.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273373
CAS Number: 129940-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR