(S)-N-Boc-allylglycine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-enoic acid, Boc-Allyl-Gly-OH, (S)-2-Boc-AMino-4-pentenoic acid, 4-Pentenoic acid, 2-[[(1,1-dimethylethoxy)carbonyl]amino]-, (2S)-

A192756-1G A192756-25G A192756-5G A192756-100MG A192756-250MG A192756-100G A192756-10G
103816-478EA 26 USD
103816-478 103847-088 77084-964 77084-966 77084-968 77192-018 77192-020 77615-632 77615-634
(S)-N-Boc-allylglycine 97%
(S)-N-Boc-allylglycine

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
Boiling Pt: 334.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01320851
CAS Number: 90600-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR