Boc-L-2-aminoadipic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)hexanedioic acid

A912649-100MG A912649-250MG A912649-1G
103841-380EA 161 USD
103841-380 76698-672 76698-674 77308-510
Boc-L-2-aminoadipic acid 95%
Boc-L-2-aminoadipic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₆
MW: 261.28 g/mol
Melting Pt: 121…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00063354
CAS Number: 77302-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR