Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Val(3-OH)-OH, 3-Hydroxy-L-valine, N-BOC protected, (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid

A182411-100MG A182411-250MG A182411-1G
103807-708EA 27 USD
103807-708 103846-080 76690-610 76690-612 77083-622
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid 97%
Boc-(S)-2-amino-3-hydroxy-3-methylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₅
MW: 233.27 g/mol
Boiling Pt: 391.1 °C (760 mm Hg)
Melting Pt: 116…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094792
CAS Number: 102507-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR