Boc-L-3,3-diphenylalanine 98%

Supplier: Ambeed
Warning

A548100-1G A548100-5G A548100-250MG
103828-676EA 42 USD
103828-676 103839-798 76843-866 76843-868 77169-694
Boc-L-3,3-diphenylalanine 98%
Boc-L-3,3-diphenylalanine

Precaución: for Research Use Only

Formula: C₂₀H₂₃NO₄
Boiling Pt: 502.7 °C (760 mm Hg)
Melting Pt: 160 °C
Flash Pt: 257.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191186
CAS Number: 138662-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR