(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid 97%

Supplier: Ambeed
Warning

A185792-250MG A185792-5G A185792-25G A185792-10G
103815-886EA 63 USD
103815-886 103846-456 76813-738 76813-740 77301-996 77671-624 77883-388
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclopentylacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
Flash Pt: 191.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05663437
CAS Number: 109183-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR