Benzyl (S)-(+)-glycidyl ether 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

A166342-10G A166342-1G A166342-5G A166342-25G A166342-100G
103806-056EA 7 USD
103806-056 103844-318 76998-040 77300-834 77300-836 77320-564 77671-344
Benzyl (S)-(+)-glycidyl ether 97%
Benzyl (S)-(+)-glycidyl ether

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Boiling Pt: 252.7 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00054428
CAS Number: 16495-13-9
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR