N-Cbz-N'-boc-L-ornithine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-(((Benzyloxy)carbonyl)amino)-5-((tert-butoxycarbonyl)amino)pentanoic acid, Nα-Benzyloxycarbonyl-Nδ-Boc-L-ornithine, Z-Orn(Boc)-OH
A835029-1G
A835029-25G
A835029-5G
A835029-100G
A835029-10G
103841-182EA
16
USD
103841-182
76845-416
76845-418
76845-420
76880-478
76880-480
N-Cbz-N'-boc-L-ornithine 98%
N-Cbz-N'-boc-L-ornithine
Precaución: for Research Use Only
Formula:
C₁₈H₂₆N₂O₆ MW: 366.414 g/mol Boiling Pt: 579.1±50.0 °C (760 mm Hg) Melting Pt: 97…103 °C Flash Pt: 304 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038268 CAS Number: 7733-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...