N-Carbobenzoxy-L-homoserine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(((Benzyloxy)carbonyl)amino)-4-hydroxybutanoic acid, Cbz-Hse-OH

A296760-1G A296760-25G A296760-10G A296760-250MG A296760-5G
103814-114EA 8 USD
103814-114 103835-256 76837-940 76837-942 76977-662 76977-664 77305-700
N-Carbobenzoxy-L-homoserine 97%
N-Carbobenzoxy-L-homoserine

Precaución: For research use only

Formula: C₁₂H₁₅NO₅
MW: 253.2512 g/mol
Boiling Pt: 519.9 °C (760 mm Hg)
Melting Pt: 100…101 °C
Flash Pt: 268.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056718
CAS Number: 35677-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR