(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Cbz-Tle-OH, Z-Tle-OH

A221267-10G A221267-500G
103818-908EA 9 USD
103818-908 103849-790 77671-966 77671-968
(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid ≥95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3,3-dimethylbutanoic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₄H₁₉NO₄
MW: 265.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00137411
CAS Number: 62965-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR