(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-enoic acid 97%

Supplier: Ambeed
Warning

A250700-100MG A250700-250MG A250700-1G A250700-5G
103811-944EA 118 USD
103811-944 103851-782 76826-938 76826-940 77091-140 77091-142
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-enoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hex-5-enoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MW: 351.4 g/mol
Boiling Pt: 573.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03094925
CAS Number: 851909-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR