(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hept-6-enoic acid 97%

Supplier: Ambeed
Warning

A210551-100MG A210551-250MG A210551-5G A210551-1G A210551-25G
103818-016EA 96 USD
103818-016 76825-872 76825-874 76825-876 77087-088 77683-598
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hept-6-enoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)hept-6-enoic acid

Precaución: For research use only.

Formula: C₂₂H₂₃NO₄
MW: 365.42 g/mol
Boiling Pt: 589.7±45.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08272862
CAS Number: 856412-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR