Fmoc-Aad(otBu)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(tert-butoxy)-6-oxohexanoic acid
A114398-100MG
A114398-10G
A114398-250MG
A114398-1G
A114398-5G
A114398-25G
103798-394EA
59
USD
103798-394
103824-234
77094-162
77094-164
77094-166
77311-046
77311-048
77900-527
Fmoc-Aad(otBu)-OH 95%
Fmoc-Aad(otBu)-OH
Precaución: for Research Use Only
Formula:
C₂₅H₂₉NO₆ Boiling Pt: 625.546 °C (760 mm Hg) Melting Pt: 108…112 °C Flash Pt: 332.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00237006 CAS Number: 159751-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...