Fmoc-β-t-butyl-L-alanine 97%
Supplier: Ambeed
|
Warning
|
A281961-100MG
A281961-10G
A281961-250MG
A281961-1G
A281961-25G
103813-582EA
66
USD
103813-582
103853-534
76802-204
76802-206
76802-208
76837-232
77305-426
Fmoc-β-t-butyl-L-alanine 97%
Fmoc-beta-t-butyl-L-alanine
Precaución: for Research Use Only
Formula:
C₂₂H₂₅NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00671389 CAS Number: 139551-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...