Fmoc-(S)-2-amino-3-methoxypropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methoxypropanoic acid, Fmoc-Ser(Me)-OH

A394173-100G A394173-250MG
103826-938EA 32 USD
103826-938 103837-976 76829-152 76829-154
Fmoc-(S)-2-amino-3-methoxypropionic acid 98%
Fmoc-(S)-2-amino-3-methoxypropionic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
MW: 341.37 g/mol
Boiling Pt: 567.9±50.0 °C (760 mm Hg)
Flash Pt: 297.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682604
CAS Number: 159610-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR