Fmoc-L-3,3-diphenylalanine 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-diphenylpropanoic acid, Fmoc-β,β-diphenyl-Ala-OH

A167210-250MG A167210-1G A167210-5G A167210-25G
103806-150EA 27 USD
103806-150 103844-412 76998-068 77081-412 77081-414 77300-882
Fmoc-L-3,3-diphenylalanine 97% (by HPLC)
Fmoc-L-3,3-diphenylalanine

Precaución: for Research Use Only

Formula: C₃₀H₂₅NO₄
MW: 463.533 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672337
CAS Number: 201484-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR