Fmoc-3-methyl-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid, Fmoc-Phe(3-Me)-OH

A150985-10G A150985-1G A150985-5G A150985-250MG
103804-224EA 22 USD
103804-224 103842-370 76997-342 77299-844 77299-846 77671-124
Fmoc-3-methyl-L-phenylalanine 97%
Fmoc-3-methyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
Boiling Pt: 628.6 °C (760 mm Hg)
Melting Pt: 130 °C
Flash Pt: 334 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863050
CAS Number: 211637-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR