N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-beta-(2-furyl)-Ala-OH, Fmoc-3-(2-Furyl)-Ala-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid, N-Fmoc-3-(2-furyl)-L-alanine

A157831-100MG A157831-10G A157831-250MG A157831-25G
103805-050EA 64 USD
103805-050 103843-254 76997-644 76997-646 76997-648 77671-236
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine

Precaución: for Research Use Only

Formula: C₂₂H₁₉NO₅
MW: 377.40 g/mol
Boiling Pt: 603.3 °C (760 mm Hg)
Melting Pt: 129.7 °C
Flash Pt: 318.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311747
CAS Number: 159611-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR