(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-fluoro-1H-indol-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A210577-100MG A210577-250MG A210577-5G A210577-1G
103818-020EA 267 USD
103818-020 77139-584 77139-586 77139-588 77303-132
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-fluoro-1H-indol-3-yl)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-fluoro-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₁FN₂O₄
MW: 444.45 g/mol
Boiling Pt: 713.9±60.0 °C (760 mm Hg)
Flash Pt: 385.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26960591
CAS Number: 1956434-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR