(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp(6-Cl)-OH

A207966-100MG A207966-250MG A207966-1G
103817-844EA 55 USD
103817-844 103848-530 76999-550 76999-552 77303-054
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₁ClN₂O₄
MW: 460.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16883068
CAS Number: 908847-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR