(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-methoxyphenyl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3-OMe)-OH

A200040-1G A200040-250MG
103817-156EA 117 USD
103817-156 103847-782 77085-842 77600-622
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-methoxyphenyl)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₅
MW: 417.45 g/mol
Boiling Pt: 647.5±55.0 °C (760 mm Hg)
Flash Pt: 345.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317009
CAS Number: 206060-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR