N-Fmoc-3-hydroxy-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-hydroxyphenyl)propanoic acid

A133756-100MG A133756-250MG A133756-5G A133756-1G
103801-914EA 56 USD
103801-914 77258-554 77258-556 77298-658 77314-600
N-Fmoc-3-hydroxy-L-phenylalanine 97%
N-Fmoc-3-hydroxy-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₅
MW: 403.427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07371961
CAS Number: 178432-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR