(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(allyloxy)phenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A547029-250MG A547029-100MG
103828-664EA 206 USD
103828-664 103839-788 76991-634 77886-362
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(allyloxy)phenyl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(allyloxy)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₅NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD08061623
CAS Number: 1175973-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR