(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,5-dichlorophenyl)propanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(2,5-DiCl)-OH

A814257-250MG A814257-5G
103841-104EA 286 USD
103841-104 77146-680 77308-370
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,5-dichlorophenyl)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,5-dichlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₉Cl₂NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD07372021
CAS Number: 1260614-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR