Fmoc-α-Me-L-Leu-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2,4-dimethylpentanoic acid, Fmoc-α-Me-Leu-OH

A154565-100MG A154565-250MG A154565-1G A154565-5G A154565-25G
103804-656EA 238 USD
103804-656 103842-846 77162-922 77162-924 77300-066 77318-580 77892-054
Fmoc-α-Me-L-Leu-OH 98%
Fmoc-alpha-Me-L-Leu-OH

Precaución: Sealed in dry, at room temperature.

Formula: C₂₂H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD08458572
CAS Number: 312624-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR