(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-cyclohexylpropanoic acid 97%

Supplier: Ambeed
Warning

A152388-100MG A152388-250MG
103804-386EA 21 USD
103804-386 103842-554 77162-862 77162-864
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-cyclohexylpropanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-cyclohexylpropanoic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₂₅H₂₉NO₄
MW: 407.5 g/mol
Boiling Pt: 588.5±29.0 °C (760 mm Hg)
Flash Pt: 309.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00156555
CAS Number: 148983-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR