H-Asp(OMe)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-tert-Butyl 4-methyl 2-aminosuccinate hydrochloride

A366278-1G A366278-5G A366278-100G A366278-10G
103826-390EA 8 USD
103826-390 103837-406 76840-642 76840-644 77551-060 77594-088
H-Asp(OMe)-OtBu·HCl 97%
H-Asp(OMe)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄·HCl
MW: 239.7 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD05662354
CAS Number: 34582-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR