(S)-1-tert-Butyl 3-methyl pyrrolidine-1,3-dicarboxylate 98%

Supplier: Ambeed
Warning

A201466-250MG A201466-1G
103817-300EA 69 USD
103817-300 103847-940 77097-404 77302-756
(S)-1-tert-Butyl 3-methyl pyrrolidine-1,3-dicarboxylate 98%
(S)-1-tert-Butyl 3-methyl pyrrolidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
Boiling Pt: 288.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09838822
CAS Number: 313706-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR