(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-butyl 3-methyl (3S)-piperazine-1,3-dicarboxylate

A137804-10G A137804-250MG A137804-1G A137804-25G A137804-5G
103802-510EA 8 USD
103802-510 77095-286 77095-288 77315-488 77315-490 77315-492
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate 97%
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.2875 g/mol
Boiling Pt: 321.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115326
CAS Number: 314741-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR