(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-tert-Butyl 2-methyl piperazine-1,2-dicarboxylate

A177962-1G A177962-5G A177962-100G A177962-100MG A177962-10G A177962-250MG A177962-25G
103807-282EA 19 USD
103807-282 103845-622 77083-018 77083-020 77138-606 77138-608 77138-610 77138-612 77301-532
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate 97%
(S)-1-tert-Butyl-2-methylpiperazine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.2875 g/mol
Boiling Pt: 321.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115328
CAS Number: 796096-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR