N-Boc-4-oxo-L-proline methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-tert-Butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, Boc-4-oxo-Pro-OMe

A228577-100G A228577-10G A228577-250MG A228577-500G A228577-1G A228577-25G A228577-5G
103819-404EA 6 USD
103819-404 103850-310 77030-980 77030-982 77030-984 77030-986 77089-104 77089-106 77089-108
N-Boc-4-oxo-L-proline methyl ester 97%
N-Boc-4-oxo-L-proline methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MW: 243.259 g/mol
Melting Pt: 42…46 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861778
CAS Number: 102195-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR