(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-tert-butyl 2-methyl (2S)-4,4-difluoropyrrolidine-1,2-dicarboxylate, (S)-1-tert-Butyl 2-methyl 4,4-difluoropyrrolidine-1,2-dicarboxylate

A131602-100MG A131602-250MG A131602-1G A131602-5G A131602-10G
103801-606EA 17 USD
103801-606 76688-644 76688-646 77314-102 77314-104 77566-552
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate 97%
(S)-1-tert-Butyl-2-methyl-4,4-difluoropyrrolidine-1,2-dicarboxylate

Precaución: For research use only.

Formula: C₁₁H₁₇F₂NO₄
MW: 265.26 g/mol
Boiling Pt: 292.7 °C (760 mm Hg)
Melting Pt: 41…46 °C
Flash Pt: 130.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094918
CAS Number: 203866-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR