(S)-(-)-1-Phenylethanol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol
A138423-100G
A138423-10G
A138423-1G
A138423-25G
A138423-5G
77137-210EA
63.85
USD
77137-210
77137-212
77137-214
77315-640
77315-642
(S)-(-)-1-Phenylethanol 98%
(S)-(-)-1-Phenylethanol
Precaución: for Research Use Only
Formula:
C₈H₁₀O Boiling Pt: 206.9 °C (760 mm Hg) Melting Pt: 9…11 °C Flash Pt: 91.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064264 CAS Number: 1445-91-6 UN: 2922 ADR: 8,6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...