(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

A236973-1G A236973-5G A236973-100G
103811-122EA 6 USD
103811-122 103850-898 77304-068 77304-070 77685-660
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98%
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Boiling Pt: 338.4 °C (760 mm Hg)
Melting Pt: 80…82 °C
Flash Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08692036
CAS Number: 118864-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR