(S)-1-N-Benzyl-prolinol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzyl-2-pyrrolidinemethanol, N-Benzyl-L-prolinol, (S)-(-)-1-Benzylpyrrolidine-2-methanol, [(2S)-1-Benzylpyrrolidin-2-yl]methanol

A294144-1G A294144-25G A294144-100G A294144-10G A294144-500G
103814-000EA 5 USD
103814-000 103835-140 76837-766 76837-768 77099-376 77099-378 77099-380
(S)-1-N-Benzyl-prolinol 97%
(S)-1-N-Benzyl-prolinol

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
MW: 191.28 g/mol
Boiling Pt: 313.3 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00183619
CAS Number: 53912-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR