(S)-3-Amino-1-methyl-piperidine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Methylpiperidin-3-amine dihydrochloride

A118276-100MG A118276-250MG
103799-024EA 33 USD
103799-024 103824-912 77161-146 77161-148
(S)-3-Amino-1-methyl-piperidine dihydrochloride 97%
(S)-3-Amino-1-methyl-piperidine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄N₂·2ClH
MW: 187.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20926142
CAS Number: 1157849-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR