(S)-1-Methyl-5-oxopyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A194175-100MG A194175-250MG
103816-628EA 63 USD
103816-628 77164-356 77892-254
(S)-1-Methyl-5-oxopyrrolidine-2-carboxylic acid 97%
(S)-1-Methyl-5-oxopyrrolidine-2-carboxylic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₆H₉NO₃
MW: 143.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19228297
CAS Number: 52574-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR