(S)-1-Boc-pyrrolidine-3-carboxylic acid 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-1-Boc-Pyrrolidine-3-carboxylic acid, (3S)-1-tert-butoxycarbonylpyrrolidine-3-carboxylic acid
A279186-100G
A279186-100MG
A279186-10G
A279186-250MG
A279186-1G
A279186-5G
A279186-25G
103813-458EA
18
USD
103813-458
103853-402
77002-112
77002-114
77002-116
77002-118
77093-336
77093-338
77305-320
(S)-1-Boc-pyrrolidine-3-carboxylic acid 97%
(S)-1-Boc-pyrrolidine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ MW: 215.25 g/mol Boiling Pt: 337.2 °C (760 mm Hg) Flash Pt: 157.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094728 CAS Number: 140148-70-5 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...