(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine 97%

Supplier: Ambeed
Warning

A132807-10G A132807-250MG A132807-1G A132807-5G
103801-784EA 61 USD
103801-784 76688-704 76688-706 77314-368 77314-370
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine 97%
(S)-(+)-1-Boc-3-(aminomethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 280.3 °C (760 mm Hg)
Flash Pt: 123.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179397
CAS Number: 199175-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR