(S)-1-Boc-(2-hydroxymethyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Boc-2-(Hydroxymethyl)piperazine, (2S)-2-(Hydroxymethyl)piperazine, N1-BOC protected

A180770-1G A180770-25G A180770-5G A180770-100MG A180770-250MG A180770-100G
103807-558EA 31 USD
103807-558 103845-920 77083-422 77083-424 77083-426 77138-706 77138-708 77685-415
(S)-1-Boc-(2-hydroxymethyl)piperazine 98%
(S)-1-Boc-(2-hydroxymethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.2800 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Flash Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772093
CAS Number: 1030377-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR