(S)-1-Boc-(3-hydroxymethyl)pyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl (3S)-3-(hydroxymethyl)pyrrolidine-1-carboxylate, (S)-1-Boc-(3-Hydroxymethyl)pyrrolidine

A227955-100G A227955-10G A227955-250MG A227955-1G A227955-25G A227955-5G
103819-348EA 23 USD
103819-348 103850-256 77013-442 77013-444 77013-446 77089-040 77089-042 77089-044
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine 98%
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Boiling Pt: 289.5 °C (760 mm Hg)
Flash Pt: 128.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094726
CAS Number: 199174-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR