(S)-(+)-1-Benzyloxy-2-propanol 96%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzyloxy-2-propanol, Benzyl (S)-(+)-2-Hydroxypropyl Ether

A100618-25G A100618-1G A100618-5G
103795-542EA 15 USD
103795-542 103821-232 77135-454 77296-780 77296-782
(S)-(+)-1-Benzyloxy-2-propanol 96%
(S)-(+)-1-Benzyloxy-2-propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 263.8±15.0 °C (760 mm Hg)
Flash Pt: 108.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07367008
CAS Number: 85483-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR