(S)-(+)-1-Aminopropan-2-ol 98%

Supplier: Ambeed
Danger

A402723-1G A402723-25G A402723-5G A402723-10G
103827-080EA 5 USD
103827-080 103838-118 76841-584 76841-586 76841-588 77003-940
(S)-(+)-1-Aminopropan-2-ol 98%
(S)-(+)-1-Aminopropan-2-ol

Precaución: for Research Use Only

Formula: C₃H₉NO
Boiling Pt: 174.5 °C (760 mm Hg)
Melting Pt: 24…26 °C
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064429
CAS Number: 2799-17-9
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR