(S)-1-Amino-3-chloropropan-2-olhydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Amino-3-chloropropan-2-ol hydrochloride

A196529-100G A196529-10G A196529-1G A196529-500G A196529-25G A196529-5G
103847-468EA 6 USD
103847-468 76986-310 76986-312 76986-314 76986-316 77085-426 77085-428
(S)-1-Amino-3-chloropropan-2-olhydrochloride 98%
(S)-1-Amino-3-chloropropan-2-olhydrochloride

Precaución: for Research Use Only

Formula: C₃H₉Cl₂NO
MW: 146.02 g/mol
Boiling Pt: 240.2 °C (760 mm Hg)
Flash Pt: 99.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056870
CAS Number: 34839-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR