(S)-1,4-Dioxane-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A406309-1G A406309-100MG A406309-250MG
103827-138EA 190 USD
103827-138 103838-176 76841-666 77155-644 77155-646
(S)-1,4-Dioxane-2-carboxylic acid 96%
(S)-1,4-Dioxane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₄
MW: 132.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26954726
CAS Number: 1821739-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR