(S)-1,2-Dimethylpiperazine 95%

Supplier: Ambeed
Danger

A195965-100MG A195965-250MG A195965-1G A195965-5G
103816-798EA 50 USD
103816-798 103847-412 76814-090 76814-092 77085-366 77085-368
(S)-1,2-Dimethylpiperazine 95%
(S)-1,2-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
Storage Temperature: Refrigerator
MDL Number: MFCD11858472
CAS Number: 485841-52-9
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR